Skip to Content
Merck
All Photos(1)

Key Documents

G1895

Sigma-Aldrich

Gly-Pro-Arg-Pro

≥95% (HPLC)

Synonym(s):

GPRP, Glycyl-Prolyl-Arginyl-Proline amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31N7O5
CAS Number:
Molecular Weight:
425.48
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

NCC(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C18H31N7O5/c19-10-14(26)24-8-2-5-12(24)15(27)23-11(4-1-7-22-18(20)21)16(28)25-9-3-6-13(25)17(29)30/h11-13H,1-10,19H2,(H,23,27)(H,29,30)(H4,20,21,22)/t11-,12-,13-/m0/s1

InChI key

WXPZDDCNKXMOMC-AVGNSLFASA-N

Gene Information

human ... FN1(2335)
mouse ... FN1(14268)
rat ... FN1(25661)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Pro-Arg-Pro

General description

Gly-Pro-Arg-Pro is a peptide that mimics the N-terminal Gly-Pro-Arg region in the α chain of fibrin protein.

Application

Gly-Pro-Arg-Pro (GPRP) has been used for the formation of crystal composed of GPRP and the 30kDa C-terminal fragment from the γ chain of fibrinogen (rFbgγC30), cloned and expressed in Pichia pastoris.

Biochem/physiol Actions

Gly-Pro-Arg-Pro (GPRP) is capable of increasing free thrombin. Thus it can be utilized to make thrombin-degranulated platelets, which helps in studying platelet reactions.
Gly-Pro-Arg-Pro peptide suppresses the early steps of fibrin polymerization. The amide derivative of this peptide prevents the degradation of fibrinogen and fragment D by plasmin, even in the presence of EGTA (ethylene glycol-bis (β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid).
Inhibits fibrin polymerization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Li et al.
European journal of haematology, 65(1), 57-65 (2000-07-29)
A flow cytometric assay for measurements of leukocyte CD11b expression in whole blood has been developed and evaluated. The method is based on triggering of the flow cytometer by a fluorescent pan leukocyte marker, RPE-CD45. This enabled flow cytometric analysis
B Shenkman et al.
The Journal of laboratory and clinical medicine, 135(1), 43-51 (2000-01-19)
Recent studies emphasize the role of blood constituents in Staphylococcus aureus (S. aureus) adherence to subendothelial extracellular matrix. In the present study, the adherence of two strains of S. aureus (ATCC 29213 and RN 6390) grown to a postexponential phase
The use of the synthetic peptide, Gly-Pro-Arg-Pro, in the preparation of thrombin-degranulated rabbit platelets
Harfenist EJ, et al.
Blood, 59(5), 952-955 (1982)
L Pasalic et al.
Journal of thrombosis and haemostasis : JTH, 16(6), 1198-1210 (2018-03-24)
Essentials Procoagulant platelets can be detected using GSAO in human whole blood. Stable coronary artery disease is associated with a heightened procoagulant platelet response. Agonist-induced procoagulant platelet response is not inhibited by aspirin alone. Collagen plus thrombin induced procoagulant platelet
Ramesh Ramapanicker et al.
Bioconjugate chemistry, 24(1), 17-25 (2012-11-16)
The synthetic tetrapeptide GPRP based on the amino-terminal GPR sequence of the fibrin α-chain binds the D-dimer protein with a dissociation constant K(D) of 25 μM. The D-dimer protein, a well-known biomarker for thrombosis, contains two cross-linked D fragments from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service