Skip to Content
Merck
All Photos(1)

Key Documents

G1046

Sigma-Aldrich

[6]-Gingerol

≥98% (HPLC)

Synonym(s):

3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (5S)-, 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, 6-Gingerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O4
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

storage condition

protect from light
under inert gas

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1

InChI

1S/C17H26O4/c1-3-4-5-6-14(18)12-15(19)9-7-13-8-10-16(20)17(11-13)21-2/h8,10-11,14,18,20H,3-7,9,12H2,1-2H3/t14-/m0/s1

InChI key

NLDDIKRKFXEWBK-AWEZNQCLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

[6]-Gingerol is implicated in apoptosis, cell cycle regulation, cytotoxic activity and inhibition of angiogenesis. It prevents serotonin release and platelet aggregation. Gingerol has immunomodulatory, antiapoptotic, antihyperglycemic, antilipidemic and antiemetic properties.

Application

[6]-Gingerol has been used:
  • to study its effects on transient receptor potential (TRP) channels
  • to study its effects on experimental models of non-alcoholic steatohepatitis
  • to determine its effects on microsomal prostaglandine E2 synthase 1 (mPGES-1), glycogen synthase kinase 3β (GSK-3β) and β-catenin pathway in A549 cell line
  • to analyse the effects of 6-Shogaol (6-SG) on diabetic nephropathy (DN) in db/db mice

Biochem/physiol Actions

Bioactive compound found in ginger (Zingiber officinale) with antioxidant activity, which functions as an anti-inflammatory and antitumor agent. [6]-Gingerol down regulates proinflammatory cytokine release by macrophages. It has been shown to inhibit COX-2 expression by blocking the activation of p38 MAP kinase and NF-κB in phorbol ester-stimulated mouse skin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[6]-Gingerol dampens hepatic steatosis and inflammation in experimental nonalcoholic steatohepatitis
Tzeng TF, et al.
Phytomedicine, 22(4), 452-461 (2015)
Yun Xu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 97, 633-641 (2017-11-05)
The prevalence of type 2 diabetes mellitus has been increasing worldwide and more than two thirds of the patients may develop diabetic nephropathy (DN). However, the efficiency of existing approaches on DN progression is limited. 6-Shogaol (6-SG), a major dehydrated
Biological properties of 6-gingerol: a brief review.
Wang S, et al.
Natural Product Communications, 9(7), 1027-1030 (2014)
Khairunnisa' Md Yusof et al.
Molecules (Basel, Switzerland), 20(6), 10280-10297 (2015-06-06)
Numerous bioactive compounds have cytotoxic properties towards cancer cells. However, most studies have used single compounds when bioactives may target different pathways and exert greater cytotoxic effects when used in combination. Therefore, the objective of this study was to determine
Effect of Herbal Supplement-Drug Interactions on Therapeutic Drug Monitoring
Chin AC and Baskin LB
Therapeutic Drug Monitoring, 417-445 (2012)

Articles

Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service