Skip to Content
Merck
All Photos(1)

Key Documents

G1019

Sigma-Aldrich

Gamborg′s Vitamin Solution

1000 ×, liquid, suitable for plant cell culture

Synonym(s):

Gamborg Vitamin Solution, Vitamin Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
10171502
NACRES:
NA.72

sterility

sterile-filtered

form

liquid

concentration

1000 ×

technique(s)

cell culture | plant: suitable

color

colorless

pH

5.0-7.0

application(s)

agriculture

storage temp.

2-8°C

Application

Gamborg′ B-5 medium described by Gamborg, et al. may be purchased as a complete medium (G5893) or reconstituted from Vitamin 1000x (G1019) and Basal Salt Mixture (G5768) chemstocks. Use Gamborg′s B-5 (B5) medium for in vitro plant culture of cell lines and tissues such as of Taxus globosa, Splachnum ampullaceum Hedw, Linum album, Eucalyptus camaldulensis. Gamborg′s B-5 medium has been used for the culture of Annona squamosa callus.
Use at a concentration of one ml per liter of prepared medium to achieve the proper final concentration.

Components

Solution contains (mg/ml): 100.0 myo-inositol, 1.0 nicotinic acid, 1.0 pyridoxine hydrochloride, 10.0 thiamine hydrochloride.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Sakamoto et al.
Plant signaling & behavior, 13(7), e1483673-e1483673 (2018-06-27)
Unrepaired DNA damage hinders the maintenance of genome integrity because it blocks the catalytic activity of replicase. The stalled replication fork can be processed through either translesion synthesis (TLS) with specific polymerases, or replication using the undamaged template. To investigate
Peng Zhao et al.
Developmental cell, 49(6), 882-893 (2019-05-14)
The maternal-to-zygotic transition (MZT) is an essential developmental turning point in both plants and animals. In plants, the timing of MZT and parental contributions to the zygotic transcriptome remain unclear. Here, by overcoming technical limitations, we characterize the Arabidopsis egg
R Mallon et al.
Cryo letters, 31(1), 24-28 (2010-03-24)
The source of germplasm as well as the technique used for storage of mosses can enhance survival after cryopreservation. Samples of gametophores, protonemata and protonemal brood cells from in vitro cultures of Splachnum ampullaceum were cryopreserved following exposure to a
Li-Da Wang et al.
Journal of Asian natural products research, 4(3), 171-174 (2002-07-18)
Callus cultures of Annona squamosa were induced using different explants including petals, seed contents (megagametophyte and embryo) and fruits (mesocarp). Growth of the calli induced from the explants was found to be influenced by the type, concentration and ratio of
Nutrient requirements of suspension cultures of soybean root cells.
O L Gamborg et al.
Experimental cell research, 50(1), 151-158 (1968-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service