F9143
Worthington LD Series Liquid Nitrogen Dewars
LD10, beaker style, capacity 10 L
Synonym(s):
cryo dewar, cryogenic storage, cryopreservation container
About This Item
Recommended Products
material
aluminum flask
Joint
Joints neck (51)
packaging
pack of 1 ea
manufacturer/tradename
Worthington 10LDB
technique(s)
cryopreservation: suitable
capacity
10 L
flask I.D. × depth
29.0 cm (11.4 in.) × 59.7 cm (23.5 in.)
neck I.D.
5.1 cm (2 in.)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
High-strength aluminum shell is both durable and light-weight. Super insulation provides advanced thermal protection. Neck tube joins inner and outer container, providing maximum durability and minimum nitrogen consumption. Cap and neck tube core provide positive closure.
Features and Benefits
- Includes a lightweight Snap-On cap for sample protection.
- Superior vacuum performance provides maximum holding times and ultimate thermal protection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service