Skip to Content
Merck

F7553

FR 122047 hydrochloride

≥98% (HPLC), powder

Synonym(s):

1-[(4,5-Bis(4-methoxyphenyl)-2-thiazoyl)carbonyl]-4-methylpiperazine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H25N3O3SHCl
CAS Number:
Molecular Weight:
459.99
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41106300
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

faintly yellow

solubility

methylene chloride: 50 mg/mL, slightly hazy, yellow

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cl[H].COc1ccc(cc1)-c2nc(sc2-c3ccc(OC)cc3)C(=O)N4CCN(C)CC4

InChI

1S/C23H25N3O3S.ClH/c1-25-12-14-26(15-13-25)23(27)22-24-20(16-4-8-18(28-2)9-5-16)21(30-22)17-6-10-19(29-3)11-7-17;/h4-11H,12-15H2,1-3H3;1H

InChI key

YWMAVHIKOAOSFM-UHFFFAOYSA-N

Biochem/physiol Actions

Selective cyclooxgenase-1 (COX-1) inhibitor.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library