Skip to Content
Merck

Skip To

F7264

Fixing solution

5 × (solution)

Synonym(s):

pre-stain fixing solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

concentration

5 × (solution)

technique(s)

protein staining: suitable

Application

An effective protein fixing solution for pre-stain fixing, suitable for SDS-PAGE, non-denaturing PAGE and IEF. Proteins require fixing into the gel for analytical purposes so that they do not leach out following electrophoresis.

Other Notes

60% (w/v) trichloroacetic acid, 17.5% (w/v) 5-sulfosalicylic acid.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D2532
concentration

5 × (solution)

concentration

-

technique(s)

protein staining: suitable

technique(s)

Southern blotting: suitable

Quality Level

200

Quality Level

300


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters