Skip to Content
Merck

F6397

IgG1−FITC Isotype Control from murine myeloma

clone MOPC 21, purified immunoglobulin, buffered aqueous solution

Synonym(s):

IgG1 Isotype Control from murine myeloma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.46
UNSPSC Code:
12352203
Conjugate:
FITC conjugate
Clone:
MOPC 21, monoclonal
Application:
FACS
Citations:
18
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

clone

MOPC 21, monoclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

flow cytometry: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3.

Application

IgG1-FITC Isotype Control from murine myeloma was used as negative control in FACS of human erythroid cell population that is expresses CD17.
IgG1?FITC Isotype Control from murine myeloma has been used in flow cytometry and as a negative staining control.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulin G (IgG) plays a major role in hypersensitivity type II and type III reactions. IgG is also involved in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


J Vollmer et al.
The Journal of investigative dermatology, 113(2), 175-181 (1999-09-01)
Nickel-induced contact dermatitis represents a T cell mediated delayed type hyperreactivity. The elucidation of the molecular basis of T cell activation by Ni2+ ions may serve as a model for the understanding of other metal allergies. We describe here the
Molecular Genetics of Immunoglobulin, 17 (1987)
Jelka Gabrilovac et al.
Immunology letters, 91(1), 39-47 (2004-02-06)
Keratinocytes actively participate in immune response and inflammation by secreting cytokines and chemokines. Membrane-bound peptidases serve as negative loop in controlling concentration of peptide signalling molecules. Recently, they have also been proposed as additional mechanism of cell-to-cell interaction and as



Global Trade Item Number

SKUGTIN
F6397-.1MG04061833618622