Skip to Content
Merck
All Photos(1)

Key Documents

F4503

Sigma-Aldrich

Ferritin from equine spleen

Type I, saline solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

sterility

sterile-filtered

Quality Level

type

Type I

form

saline solution

mol wt

H subunit ~21 kDa
L subunit ~19 kDa

color

red to brown

cation traces

Cd: ≤1% (as % of ferritin)

storage temp.

2-8°C

General description

A shell of 24 protein subunits (apoferritin) and a core Fe 3+ ions with a peptide MW of 440 kDa. Review of the ferritin-like superfamily of iron-storage proteins, virtually ubiquitous in life on earth.

Application

Ferritin from equine spleen has been used:
  • in mycobacterial growth assay
  • in size-exclusion chromatography
  • in labeling of macrophage cells
  • as a reference standard in muon spin spectroscopy

Biochem/physiol Actions

A ubiquitous iron storage protein that plays a key role in iron metabolism. It serves as an intracellular iron reserve (particularly in spleen, liver, intestinal mucosa, and bone marrow) and functions in iron detoxification.
A ubiquitous iron storage protein that plays a key role in iron metabolism. It serves as an intracellular iron reserve (particularly in spleen, liver, intestinal mucosa, and bone marrow) and functions in iron detoxification. Studies have shown that ferritin iron incorporation is mediated by a ferroxidase activity associated with ferritin H subunits and a nucleation center associated with ferritin L subunits. Release of iron from ferritin has an essential role in iron-dependent lipid peroxidation and may contribute to free radical-induced cell damage in vivo. Therefore, by binding iron, ferritin may function as an antioxidant.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human-brain ferritin studied by muon spin rotation: a pilot study
Bossoni L, et al.
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 29(41), 415801-415801 (2017)
Italo Moglia et al.
Journal of inorganic biochemistry, 183, 184-190 (2017-12-28)
Ferritin is a globular protein that consists of 24 subunits forming a hollow nanocage structure that naturally stores iron oxyhydroxides. Elimination of iron atoms to obtain the empty protein called apoferritin is the first step to use this organic shell
The effectiveness of ferritin as a contrast agent for cell tracking MRI in mouse cancer models
Lee CW, et al.
Yonsei Medical Journal, 58(1), 51-58 (2017)
Yves Gossuin et al.
Magnetic resonance in medicine, 48(6), 959-964 (2002-12-05)
Ferritin, the iron-storing protein, speeds up proton transverse magnetic relaxation in aqueous solutions. This T(2) shortening is used in MRI to quantify iron in the brain and liver. Current theoretical models underestimate the relaxation enhancement by ferritin at imaging fields
Adipocyte model of Mycobacterium tuberculosis infection reveals differential iron availability to bacilli in the lipid rich caseous environment
Nandy A, et al.
Infection and Immunity, IAI-00041 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service