Skip to Content
Merck
All Photos(1)

Documents

F2012

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:64 (minimum)

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein and a major class of immunoglobulin. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.
The product binds all mouse Igs.

Application

Anti-Mouse IgG (whole molecule)−FITC antibody has been used:
  • in immunofluorescence
  • in immunohistochemistry
  • in immunofluorescence staining
  • in immunofluorescence microscopy
  • in confocal imaging

Anti-Mouse IgG (whole molecule)-FITC antibody produced in goat was used for FACS analysis of DF3 antigen on breast cancer cell lines.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of FITC to Anti-Mouse IgG (whole molecule) antibody allows for the visualization of protein by fluorescent microscopy.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with human samples

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
The Laboratory Rat (2005)
Adhesion and differentiation of human mesenchymal stem cells on plasma-functionalized graphenes with different feeding gases
Baik K Y, et al.
Carbon, 77(10), 302-310 (2014)
L Chen et al.
The Journal of clinical investigation, 96(6), 2775-2782 (1995-12-01)
The high molecular weight mucin-like glycoprotein, DF3 (MUC1), is overexpressed in the majority of human breast cancers. Here we demonstrate that replication defective recombinant adenoviral vectors, containing the DF3 promoter (bp -725 to +31), can be used to express beta-galactosidase
Hemoglobin subunit beta interacts with the capsid protein and antagonizes the growth of classical swine fever virus
Li D, et al.
Journal of virology, 414(2-3), JVI-03130 (2013)
Exposure to ELF-pulse modulated X band microwaves increases in vitro human astrocytoma cell proliferation
Perez-Castejon C, et al.
Histology and Histopathology, 24(10), 1551-1551 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service