Skip to Content
Merck
All Photos(2)

Key Documents

F0638

Sigma-Aldrich

Iron(III) sulfate hydrate

BioReagent, suitable for plant cell culture

Synonym(s):

Ferric sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
grade:
reagent
form:
powder

grade

reagent

product line

BioReagent

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Fe.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

RUTXIHLAWFEWGM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Iron(III) sulfate hydrate is a general use reagent qualified for use in plant research. It is redox active and my be used as a cell culture nutrient, in studies on iron metabolism or regulation and oxidative processes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Franza et al.
Molecular plant-microbe interactions : MPMI, 12(2), 119-128 (1999-02-02)
Low iron availability is a triggering signal for coordinated expression of the genes encoding pectate lyases PelB, PelC, PelD, and PelE, and chrysobactin iron transport functions, which are two main determinants of phytopathogenicity of the Erwinia chrysanthemi strain 3937. The
Neeraja Vajrala et al.
BMC microbiology, 11, 37-37 (2011-02-23)
In response to environmental iron concentrations, many bacteria coordinately regulate transcription of genes involved in iron acquisition via the ferric uptake regulation (Fur) system. The genome of Nitrosomonas europaea, an ammonia-oxidizing bacterium, carries three genes (NE0616, NE0730 and NE1722) encoding
Luis A Sayavedra-Soto et al.
Methods in enzymology, 486, 403-428 (2010-12-28)
The chemolithoautotroph Nitrosomonas europaea oxidizes about 25 mol of NH(3) for each mole of CO(2) that is converted to biomass using an array of heme and nonheme Fe-containing proteins. Hence mechanisms of efficient iron (Fe) uptake and homeostasis are particularly
Jaita Pal et al.
Indian journal of experimental biology, 48(12), 1210-1218 (2011-01-22)
Cellular damage caused by reactive oxygen species has been implicated in several diseases and hence antioxidants have significant importance in human health. Cold water, hot water and methanolic extract of Pleurotus squarrosulus were evaluated for antioxidant activity against hydroxyl radical
Ricardo Arevalo et al.
Rapid communications in mass spectrometry : RCM, 32(21), 1875-1886 (2018-07-27)
The investigation of cryogenic planetary environments as potential harbors for extant life and/or contemporary sites of organic synthesis represents an emerging focal point in planetary exploration. Next generation instruments need to be capable of unambiguously determining elemental and/or molecular stoichiometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service