Skip to Content
Merck
All Photos(1)

Key Documents

EP0030129326

Eppendorf® Twin tec microbiology PCR plates, 96 wells

colorless plate, 96, semi-skirted, conical bottom colorless wells, PCR clean, semi-skirted, pkg of 10 ea, colorless

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

Pricing and availability is not currently available.

material

colorless plate
conical bottom colorless wells

feature

PCR clean
skirt
96, semi-skirted

packaging

pkg of 10 ea

manufacturer/tradename

Eppendorf® 0030129326

size

96 wells

color

colorless

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SML0248SML1926SML1886
ZCZ011 ≥98% (HPLC)

SML2710

ZCZ011

GAT211 ≥98% (HPLC)

SML1926

GAT211

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: ≥5 mg/mL

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service