Key Documents
EP0030128460
Eppendorf® Twin tec PCR plates, 96 wells
skirted, with barcode, individual color (wells colorless), pkg of 25 ea
About This Item
UNSPSC Code:
41106300
feature
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128460
technique(s)
PCR: suitable
size
96 wells
General description
Choose the Eppendorf twin.tec® polymerase chain reaction (PCR) plates for confidence in every PCR assay. Twin.tec® plates offer purity, rigidity, color, and other simple, smart features that can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow. All Eppendorf twin.tec® PCR plates are ‘PCR clean’. With twin.tec® PCR plates you’ll have maximum confidence in your results.
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Application
Eppendorf® Twin tec® PCR plates, 96 wells are suitable for use in:
  • standard PCR
  • real-time PCR
  • high throughput PCR
  • small volume sample handling
Features and Benefits
  • The design of the product is unified, with a polycarbonate frame and polypropylene wells, ensuring optimal performance.
  • The polypropylene wells are very thin walled, allowing for efficient heat transfer to the sample.
  • The polycarbonate frame is exceptionally sturdy and resistant to torque.
  • Enhanced well-to-well tolerance.
  • Incorporated cut-away corners and alphanumeric grid-referencing.
  • Elevated well rims facilitate effective sealing, minimizing the risk of cross-contamination.
  • Batch-tested and certified to be DNA, DNase, RNase, and PCR inhibitors free (PCR clean).
  • Optional barcode available upon request for skirted and semi-skirted versions.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG