EP0030127838
Eppendorf® heat sealing film
PCR clean, pack of 100 ea
Synonym(s):
Eppendorf® Heat Sealing Film
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.15
Recommended Products
material
clear polyester
sterility
non-sterile
feature
PCR clean
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 0030127838
technique(s)
PCR: suitable
suitability
suitable for PCR
General description
Panels can be effectively sealed with PCR Films & Foils.Eppendorf PCR Films and Foils provide an effective and airtight plate seal to better protect PCR samples during temperature changes and storage. PCR sealing films have excellent chemical resistance and have been tested for their resistance to a selection of common laboratory substances.
Application
PCR
Features and Benefits
- Hermetic sealing of multiwell plates, especially recommended for low reaction volumes
- Best protection against evaporation during PCR
- Maximum adhesiveness at high temperatures during PCR
- Adhesive flaps help to position the film for sealing
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service