Skip to Content
Merck
All Photos(1)

Key Documents

EMU080691

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Taf8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAGAGCAAGATGGCTGACACGGCGGCCGGGCCTGGTGGCTCCGGAACGAGACCGGGAAGTAAGCAATCCACTAACCCTGCTGATAACTACCATCTGGCCCGGAGGAGGACCCTGCAAGTGGTTGTGAGCTCCTTGCTGACAGAGGCGGGATTTGAGAGTGCCGAGAAAGCGTCTGTGGAGACACTGACAGAGATGCTGCAGAGCTACATCTCAGAAATCGGGAGAAGTGCTAAGTCATACTGTGAGCACACAGCCAGGACCCAGCCCACACTGTCGGACATCGTGGTCACATTGGTTGAAATGGGCTTCAATGTTGACACTCTACCTGCCTATGCAAAACGGTCTCAGAGGATGGTCATCACTGCACCTCCAGTGACCAATCAGCCAGTGACCCCCAAGGCCCTCACTGCAGGGCAGAACCGGCCCCACCCGCCACACATCCCCAGCCATTTTCCTGAGTTTCCGGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuya Muto et al.
Investigative ophthalmology & visual science, 55(7), 4327-4337 (2014-06-19)
To investigate whether high glucose (HG) alters connexin 43 (Cx43) expression and gap junction intercellular communication (GJIC) activity in retinal Müller cells, and promotes Müller cell and pericyte loss. Retinal Müller cells (rMC-1) and cocultures of rMC-1 and retinal pericytes
Caitlin J Bowen et al.
Developmental biology, 407(1), 145-157 (2015-07-19)
Proper remodeling of the endocardial cushions into thin fibrous valves is essential for gestational progression and long-term function. This process involves dynamic interactions between resident cells and their local environment, much of which is not understood. In this study, we
Tomas Baldassarre et al.
Molecular cancer research : MCR, 13(6), 1044-1055 (2015-03-19)
Triple-negative breast cancers (TNBCs) are highly aggressive cancers that lack targeted therapies. However, EGFR is frequently activated in a subset of TNBCs and represents a viable clinical target. Because the endocytic adaptor protein Endophilin A2 (SH3GL1/Endo II) has been implicated
Zheren Shao et al.
PloS one, 10(7), e0132655-e0132655 (2015-07-24)
Melanomas cause over 76% of skin cancer deaths annually. Phosphatidylinositol 3-kinase (PI3K)-AKT-mammalian target of rapamycin (mTOR) signaling pathway is important for melanoma initiation and progression. In the current study, we evaluated the potential anti-melanoma effect of VS-5584, a novel and
Alexandre K Rouquette-Jazdanian et al.
PloS one, 10(6), e0131823-e0131823 (2015-06-30)
Linker for Activation of T cells (LAT) is an adapter protein that is essential for T cell function. Knock-in mice with a LAT mutation impairing calcium flux develop a fatal CD4+ lymphoproliferative disease. miR-155 is a microRNA that is correlated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service