Skip to Content
Merck
All Photos(1)

Documents

EHU065921

Sigma-Aldrich

MISSION® esiRNA

targeting human PRMT5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCAGTGGCTCTTGAAATTGGGGCTGACCTCCCATCTAATCATGTCATTGATCGCTGGCTTGGGGAGCCCATCAAAGCAGCCATTCTCCCCACTAGCATTTTCCTGACCAATAAGAAGGGATTTCCTGTTCTTTCTAAGATGCACCAGAGGCTCATCTTCCGGCTCCTCAAGTTGGAGGTGCAGTTCATCATCACAGGCACCAACCACCACTCAGAGAAGGAGTTCTGCTCCTACCTCCAATACCTGGAATACTTAAGCCAGAACCGTCCTCCACCTAATGCCTATGAACTCTTTGCCAAGGGCTATGAAGACTATCTGCAGTCCCCGCTTCAGCCACTGATGGACAATCTGGAATCTCAGACATATGAAGTGTTTGAAAAGGACCCCATCAAATACTCTCAGTACCAGCAGGCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Biochem/physiol Actions

PRMTs (protein arginine N-methyltransferases) are responsible for the methylation of arginine residues in histones. PRMT5 causes monomethylation and symmetric dimethylation reactions. It works as an oncogene is some neoplasms and is associated with cell proliferation, inhibition of cell death and regulation of tumor suppressor genes. PRMT5 is also involved in stem cell maintenance by controlling differentiation-associated genes. It is involved in cellular responses, including neurogenesis and myogenesis metabolic events, somatic cell reprogramming, regulation of Golgi apparatus, ribosome biogenesis and neuronal spreading and movements.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ju-Yeon Jeon et al.
Oncology reports, 40(1), 536-544 (2018-05-12)
Protein arginine methyltransferase 5 (PRMT5) is a protein that catalyzes transfer of methyl groups to the arginine residues of proteins and is involved in diverse cellular and biological responses. While the participation of PRMT5 in cancer progression has been increasingly documented
Nan Wang et al.
Cell death & disease, 11(10), 864-864 (2020-10-17)
Metastasis is the main cause of laryngeal cancer-related death; its molecular mechanism remains unknown. Here we identify protein arginine methyltransferase 5 (PRMT5) as a new metastasis-promoting factor in laryngeal carcinoma, and explore its underlying mechanism of action in regulating laryngeal
Dongying Chen et al.
Journal of cellular and molecular medicine, 21(4), 781-790 (2016-11-20)
To probe the role of protein arginine methyltransferase 5 (PRMT5) in regulating inflammation, cell proliferation, migration and invasion of fibroblast-like synoviocytes (FLSs) from patients with rheumatoid arthritis (RA). FLSs were separated from synovial tissues (STs) from patients with RA and
Myc and Omomyc functionally associate with the Protein Arginine Methyltransferase 5 (PRMT5) in glioblastoma cells.
Mongiardi MP
Scientific Reports, 5, 15494-15494 (2015)
Shikui Zhang et al.
Journal of cellular and molecular medicine, 23(2), 1333-1342 (2018-11-22)
The emerging evidence reveals that protein arginine methyltransferase 5 (PRMT5) is involved in regulation of tumour cell proliferation and cancer development. Nevertheless, the exact role of PRMT5 in human lung cancer cell proliferation and the underlying molecular mechanism remains largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service