Skip to Content
Merck
All Photos(1)

Documents

EHU065071

Sigma-Aldrich

MISSION® esiRNA

targeting human ITGB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGGCTTTACGGAGGAAGTAGAGGTTATTCTTCAGTACATCTGTGAATGTGAATGCCAAAGCGAAGGCATCCCTGAAAGTCCCAAGTGTCATGAAGGAAATGGGACATTTGAGTGTGGCGCGTGCAGGTGCAATGAAGGGCGTGTTGGTAGACATTGTGAATGCAGCACAGATGAAGTTAACAGTGAAGACATGGATGCTTACTGCAGGAAAGAAAACAGTTCAGAAATCTGCAGTAACAATGGAGAGTGCGTCTGCGGACAGTGTGTTTGTAGGAAGAGGGATAATACAAATGAAATTTATTCTGGCAAATTCTGCGAGTGTGATAATTTCAACTGTGATAGATCCAATGGCTTAATTTGTGGAGGAAATGGTGTTTGCAAGTGTCGTGTGTGTGAGTGCAACCCCAACTACACTGGCAGTGCATGTGACTGTTCTTTGGATACTAGTACTTGTGAAGCCAGCAACG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen Wantoch von Rekowski et al.
Biomolecules, 9(12) (2019-11-30)
Tumor cell binding to the microenvironment is regarded as the onset of therapeutic resistance, referred to as cell adhesion mediated drug resistance (CAM-DR). Here we elucidate whether CAM-DR occurs in ovarian cancer cells and contributes to still-existing cisplatin resistance. Cultivation
Wenjie Yang et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(1), 132-144 (2020-02-13)
This study aimed to investigate the regulatory mechanism of circular RNA CSPP1 (hsa_circ_CSPP1) in cervical cancer. Based on GEO database, differentially expressed circRNAs and mRNAs related to cervical cancer were screened out by R software. Kyoto Encyclopedia of Genes and
Zahia Hamidouche et al.
BMC cell biology, 11, 44-44 (2010-06-25)
The potential of mesenchymal stromal cells (MSCs) to differentiate into functional bone forming cells provides an important tool for bone regeneration. The identification of factors that trigger osteoblast differentiation in MSCs is therefore critical to promote the osteogenic potential of
Xiao-Min Wang et al.
PloS one, 8(2), e55714-e55714 (2013-02-27)
To explore the key regulatory genes associated with lung cancer in order to reduce its occurrence and progress through silencing these key genes. To identify the key regulatory genes involved in lung cancer, we performed a combination of gene array
Luisa Boldrin et al.
PLoS currents, 3, RRN1294-RRN1294 (2012-02-16)
Satellite cells, normally quiescent underneath the myofibre basal lamina, are skeletal muscle stem cells responsible for postnatal muscle growth, repair and regeneration. Since their scarcity and small size have limited study on transverse muscle sections, techniques to isolate individual myofibres

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service