Skip to Content
Merck
All Photos(1)

Key Documents

EHU062481

Sigma-Aldrich

MISSION® esiRNA

targeting human MAPK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTGGAAGCCATGAGAGATGTCTACATTGTGCAGGACCTGATGGAGACTGACCTGTACAAGTTGCTGAAAAGCCAGCAGCTGAGCAATGACCATATCTGCTACTTCCTCTACCAGATCCTGCGGGGCCTCAAGTACATCCACTCCGCCAACGTGCTCCACCGAGATCTAAAGCCCTCCAACCTGCTCATCAACACCACCTGCGACCTTAAGATTTGTGATTTCGGCCTGGCCCGGATTGCCGATCCTGAGCATGACCACACCGGCTTCCTGACGGAGTATGTGGCTACGCGCTGGTACCGGGCCCCAGAGATCATGCTGAACTCCAAGGGCTATACCAAGTCCATCGACATCTGGTCTGTGGGCTGCATTCTGGCTGAGATGCTCTCTAACCGGCCCATCTTCCCTGGCAAGCACTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Ricard et al.
Cells, 9(1) (2019-12-28)
Despite the clinical importance of arteriogenesis, this biological process is poorly understood. ERK1 and ERK2 are key components of a major intracellular signaling pathway activated by vascular endothelial growth (VEGF) and FGF2, growth factors critical to arteriogenesis. To investigate the
Osamu Otabe et al.
Oncology reports, 37(1), 98-104 (2016-11-15)
In alveolar rhabdomyosarcoma (ARMS) that is a highly malignant pediatric soft tissue tumor, MET, a receptor of hepatocyte growth factor (HGF), was reported to be downstream of the PAX3-FOXO1 fusion gene specific to ARMS, and a key mediator of metastatic
Yunching Chen et al.
Scientific reports, 7, 44123-44123 (2017-03-10)
Sorafenib is a RAF inhibitor approved for several cancers, including hepatocellular carcinoma (HCC). Inhibition of RAF kinases can induce a dose-dependent "paradoxical" upregulation of the downstream mitogen-activated protein kinase (MAPK) pathway in cancer cells. It is unknown whether "paradoxical" ERK
Wei Chen et al.
Cell & bioscience, 7, 43-43 (2017-08-31)
Connexins are a family of transmembrane proteins that form gap junctions, which are important for diffusion of cytosolic factors such as ions and second messenger signaling molecules. Our previous study has shown that Connexin40 (Cx40), one dominant connexin expressed in
Namal Perera et al.
International journal of biological sciences, 14(10), 1378-1388 (2018-08-21)
Antrodia cinnamomea (A. cinnamomea) is a medicinal fungus used in traditional Chinese medicine to treat different kinds of ailments, including liver diseases, abdominal pain, drug intoxication, diarrhea, itchy skin, hypertension, and cancer. Polysaccharides have been identified as one of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service