Skip to Content
Merck
All Photos(1)

Key Documents

EHU002721

Sigma-Aldrich

MISSION® esiRNA

targeting human ERN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACAGTGACGCTTCCTGAAACCTTGTTGTTTGTGTCAACGCTGGATGGAAGTTTGCATGCTGTCAGCAAGAGGACAGGCTCAATCAAATGGACTTTAAAAGAAGATCCAGTCCTGCAGGTCCCAACACATGTGGAAGAGCCTGCCTTTCTCCCAGATCCTAATGATGGCAGCCTGTATACGCTTGGAAGCAAGAATAATGAAGGCCTGACGAAACTTCCTTTTACCATCCCAGAATTGGTGCAGGCATCCCCATGCCGAAGTTCAGATGGAATCCTCTACATGGGTAAAAAGCAGGACATCTGGTATGTTATTGACCTCCTGACCGGAGAGAAGCAGCAGACTTTGTCATCGGCCTTTGCAGATAGTCTCTGCCCATCAACCTCTCTTCTGTATCTTGGGCGAACAGAATACACCATCACCATGTACGACACCAAAACCCGAGAGCTCCGGTGGAATGCCACCTACTTTGACTATGCGGCCTCACTGCCTGAGGACGACGTGGACTACAAGATGTCCCACTTTGTGTCCAATGGTGATGGGCTGGTGGTGACTGTGGACAGT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Sophie Michallet et al.
PloS one, 6(10), e25820-e25820 (2011-10-27)
To determine whether the Unfolded Protein Response (UPR) sensors (PERK, ATF6 and IRE-1) can be targeted to promote death of Multiple Myeloma (MM) cells. We have knocked-down separately each UPR stress sensor in human MM cell lines using RNA interference
Qun Lai et al.
Yonsei medical journal, 54(6), 1407-1415 (2013-10-22)
To investigate the anti-apoptotic mechanism of leptin in non-small cell lung cancer. The influences of leptin on apoptosis were investigated, analyzing the mechanism that triggers growth of A549 cells. The effects of leptin on cell proliferation were examined by XTT
Xia Sheng et al.
EMBO molecular medicine, 7(6), 788-801 (2015-04-13)
The unfolded protein response (UPR) is a homeostatic mechanism to maintain endoplasmic reticulum (ER) function. The UPR is activated by various physiological conditions as well as in disease states, such as cancer. As androgens regulate secretion and development of the
K W Kim et al.
Oncogene, 29(22), 3241-3251 (2010-03-30)
As apoptosis defects limit efficacy of anticancer agents, autophagy has been proposed as a novel strategy for radiotherapy enhancement. We previously showed that caspase-3/7 inhibition induces autophagy and promotes radiosensitivity in vitro and in vivo. Therefore, we further investigated the
Amanda Crider et al.
Molecular neurobiology, 55(9), 7606-7618 (2018-02-13)
Impaired social interaction is a key feature of several major psychiatric disorders including depression, autism, and schizophrenia. While, anatomically, the prefrontal cortex (PFC) is known as a key regulator of social behavior, little is known about the cellular mechanisms that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service