Skip to Content
Merck
All Photos(2)

Key Documents

E7659

Sigma-Aldrich

Anti-EEA1 antibody, Mouse monoclonal

clone EEA1-N19, purified from hybridoma cell culture

Synonym(s):

Anti-MST105, Anti-MSTP105, Anti-ZFYVE2, Monoclonal Anti-Early endosomal antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

EEA1-N19, monoclonal

form

buffered aqueous solution

mol wt

antigen ~160 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using whole extract of rat NRK cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EEA1(8411)
mouse ... Eea1(216238)
rat ... Eea1(314764)

General description

Monoclonal Anti-EEA1 (mouse IgG2a isotype) is derived from the hybridoma EEA1-N19 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of human EEA1. Early Endosome Antigen 1 (EEA1) is a dimer. It comprises extensive coiled-coil regions, and at its C-terminus contains a cysteine-rich zinc-finger-like domain named FYVE domain.
The gene Early Endosome Antigen 1 (EEA1) encodes for around 1400 amino acid proteins. It is a peripheral membrane protein associated with the cytoplasmic face of early endosomes. EEA1 is a autoantigen associated with subacute cutaneous systemic lupus erythematosus and is a coiled-coil protein localized to early endosomes and cytosol.

Application

Anti-EEA1 antibody, Mouse monoclonalhas been used in:
  • immunoblotting
  • immunoprecipitation
  • cell fractionation
  • indirect immunofluorescence
  • immunohistochemistry

Monoclonal Anti-EEA1 antibody produced in mouse is suitable for the following applications:
  • Immunoprecipitation
  • Indirect immunofluorescence
  • Western blotting at a concentration of 1-2μg/mL using whole extract of rat NRK cells

Biochem/physiol Actions

Early Endosome Antigen 1 (EEA1) FYVE domain is involved in intracellular trafficking and is implicated in the specific localization of EEA1 to endosomes . Endosomal targeting of EEA1 also requires its binding to the active form of the small GTPase Rabaptin-5 (Rab5). The binding of EEA1 to phosphatidylinositol 3-phosphate (PtdIns 3P) and Rab5-GTP is essential for the localization and function of EEA1 in endocytic membrane fusion. Monoclonal Anti-EEA1 may be used as an early endosome marker.
Early Endosome Antigen 1 (EEA1) controls vesicle fusion during endocytosis. In neurons, it is involved in recycling of synaptic vesicles and neurotransmitter receptors. Patients with neurological deficits develop EEA1 autoantibodies. Mutation in EEA1 gene leads to susceptibility to diabetes in the Japanese population. Phosphatidylinositol 3-phosphate (PtdIns(3)P) is essential for the localization and function of EEA1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Timely regulated sorting from early to late endosomes is required to maintain cerebellar long-term depression
Kim T, et al.
Nature Communications, 8(1), 401-401 (2017)
Cellular functions of phosphatidylinositol 3-phosphate and FYVE domain proteins
GILLOOLY DJ, et al.
The Biochemical Journal, 355, 249-258 (2001)
Biogenesis of the sorting endosome: the role of Rab5
Woodman PG
Traffic, 1(9), 695-701 (2000)
Taegon Kim et al.
Nature communications, 8(1), 401-401 (2017-09-03)
An important feature of long-term synaptic plasticity is the prolonged maintenance of plastic changes in synaptic transmission. The trafficking of AMPA-type glutamate receptors (AMPARs) is involved in the expression of many forms of synaptic plasticity, yet the subsequent events accomplishing
Anna A Cook et al.
eLife, 12 (2023-12-12)
Spinocerebellar ataxia type 6 (SCA6) is a rare disease that is characterized by cerebellar dysfunction. Patients have progressive motor coordination impairment, and postmortem brain tissue reveals degeneration of cerebellar Purkinje cells and a reduced level of cerebellar brain-derived neurotrophic factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service