Skip to Content
Merck
All Photos(3)

Key Documents

E6760

Sigma-Aldrich

Ethylenediaminetetraacetic acid ferric sodium salt

BioReagent, suitable for plant cell culture, crystalline

Synonym(s):

Ethylenediaminetetraacetic acid iron(III) sodium salt, Ferric sodium EDTA, EDTA iron(III) sodium salt, Edathamil, Komplexon II

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12N2NaFeO8
CAS Number:
Molecular Weight:
367.05
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.85

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

crystalline

technique(s)

cell culture | plant: suitable

storage temp.

room temp

SMILES string

[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Fe.Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;+3;+1/p-4

InChI key

MKWYFZFMAMBPQK-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. Iron-EDTA enhances damage due to reactive oxygen species, such as depletion of reduced glutathione, formation of methemoglobin, and lipid peroxidation.

Biochem/physiol Actions

Chelates divalent cations.

Legal Information

Komplexon is a trademark of CU Chemie Uetikon AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B B Christensen et al.
Gene, 173(1 Spec No), 59-65 (1996-01-01)
Horizontal transfer of the TOL plasmid was examined in Pseudomonas putida (Pp) KT2442 micro-colonies on semi-solid agar surfaces. Horizontal gene transfer is usually studied in large populations where all information is based on average estimates of the transfer events in
Prem L Bhalla et al.
Nature protocols, 3(2), 181-189 (2008-02-16)
Agrobacterium-mediated transformation is widely used for gene delivery in plants. However, commercial cultivars of crop plants are often recalcitrant to transformation because the protocols established for model varieties are not directly applicable to them. The genus Brassica includes the oil
Wei Xia et al.
The British journal of nutrition, 108(2), 357-362 (2011-10-19)
In the present study, we investigated the prevalence of anaemia and Fe deficiency anaemia (IDA) and explored the relationship between Helicobacter pylori infection and IDA in adolescent girls. A total of 1037 adolescent girls from Suihua, China were enrolled. Hb
Zuoming Zhou et al.
Bioresource technology, 132, 210-216 (2013-02-16)
Biological reduction of Fe(III) to Fe(II) is a key step in nitrogen oxides (NOx) removal by the integrated chemical absorption-biological reduction method, which determines the concentration of Fe(II) in the scrubbing liquid. A new Fe(III)EDTA reduction strain, named as FD-3
Bo Wang et al.
Wei sheng yan jiu = Journal of hygiene research, 40(3), 334-337 (2011-06-24)
To assess the effectiveness of social mobilization and social marketing in promoting NaFeEDTA-fortified soy sauce in an iron deficient population. This study was an uncontrolled, community-based, before-after study, which was implemented in three counties of Shijiazhuang Municipality. The intervention was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service