Skip to Content
Merck

E5280

pT7-FLAG-MAT-Tag®-1 Expression Vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
12171500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

Quality Segment

storage temp.

−20°C

Application

Cytoplasmic expression of Met-N-terminal FLAG®, C-terminal MAT-Tag® dual tagged fusion proteins under the T7/lac promoter.

Other Notes

Browse additional application references in our FLAG® Literature portal.
Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG is a trademark of Sigma-Aldrich Co. LLC
pT7-FLAG-MAT is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
E5280-10UG04061835518104