Skip to Content
Merck
All Photos(1)

Key Documents

E1641

Sigma-Aldrich

Epibrassinolide

≥85%

Synonym(s):

22R,23R,24R-2α,3α,22,23-Tetrahydroxy-B-homo-7-oxa-5α-ergostan-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48O6
CAS Number:
Molecular Weight:
480.68
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥85%

application(s)

agriculture

storage temp.

−20°C

SMILES string

[H][C@@]1(CC[C@@]2([H])[C@]3([H])COC(=O)[C@@]4([H])C[C@H](O)[C@H](O)C[C@]4(C)[C@@]3([H])CC[C@]12C)[C@H](C)[C@@H](O)[C@H](O)[C@H](C)C(C)C

InChI

1S/C28H48O6/c1-14(2)15(3)24(31)25(32)16(4)18-7-8-19-17-13-34-26(33)21-11-22(29)23(30)12-28(21,6)20(17)9-10-27(18,19)5/h14-25,29-32H,7-13H2,1-6H3/t15-,16+,17+,18-,19+,20+,21-,22+,23-,24-,25-,27-,28-/m1/s1

InChI key

IXVMHGVQKLDRKH-QHBHMFGVSA-N

Application

Epibrassinolide has been used:
  • in Murashige and Skoog (MS) medium to study the cotyledon and shoot length response of Cappelle-Desprez (rht8) and RIL6 (Rht8) plants towards it
  • in callus-inducing medium for callus induction
  • to induce the expression of the gene leaf inclination 2 (LC2) in rice sampling

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Sadava et al.
Biochemical and biophysical research communications, 493(1), 783-787 (2017-08-30)
Small-cell lung carcinoma (SCLC) has a dismal prognosis in part because of multidrug resistance (MDR). Epibrassinolide (EB) is a steroid hormone in plants, with many physiological effects. It acts via a membrane receptor and GSK3 pathway, resulting in stabilization of
Jiangmei Sun et al.
BMC genomics, 18(1), 763-763 (2017-10-12)
Leucine-rich repeat receptor-like protein kinase (LRR-RLK) is the largest gene family of receptor-like protein kinases (RLKs) and actively participates in regulating the growth, development, signal transduction, immunity, and stress responses of plants. However, the patterns of LRR-RLK gene family evolution
RNA-Seq analysis of the Arabidopsis transcriptome in pluripotent calli
Lee K, et al.
Molecules and Cells, 39(6), 484-484 (2016)
W A Stirk et al.
Plant physiology and biochemistry : PPB, 70, 348-353 (2013-07-03)
Endogenous gibberellins and brassinosteroids were quantified in 24 axenic microalgae strains from the Chlorophyceae, Trebouxiophyceae, Ulvophyceae and Charophyceae microalgae strains after 4 days in culture. This is the first report of endogenous gibberellins being successfully detected in microalgae. Between 18
Yuqing He et al.
The New phytologist, 214(1), 388-399 (2016-12-16)
Plant hormones play a vital role in plant immune responses. However, in contrast to the relative wealth of information on hormone-mediated immunity in dicot plants, little information is available on monocot-virus defense systems. We used a high-throughput-sequencing approach to compare

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service