Skip to Content
Merck

E1005

Electrophoresis film for agarose gels

roll W × L 5 in. × 54 ft

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41116155
Material:
polyester
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyester

description

Film 7 mil (0.2 mm) Thickness

roll W × L

5 in. × 54 ft

General description

  • Gels cast on the electrophoresis film remain permanently attached to the film during electrophoresis, staining and drying
  • Handling of thin agarose gels is much easier when attached to electrophoresis film
  • Gels attached to electrophoresis film remain flexible after drying

Application

Electrophoresis film is a flexible, transparent polyester film specifically designed to support agarose gels. Ideal for casting horizontal agarose gels for immunoelectrophoresis and immunodiffusion. One side has been treated with a hydrophilic coating, which enhances adhesion with aqueous agarose gel media.

Disclaimer

Fluorescence may be exhibited under ultraviolet light which may interfere with some fluorimetric detection methods.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library