Skip to Content
Merck
All Photos(1)

Key Documents

D8940

Sigma-Aldrich

Anti-Destrin/ADF (AE-14) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Actin Depolymerizing Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~0.8-1.2 mg/mL

technique(s)

microarray: suitable
western blot: 1:600 using whole extracts of human A431 epidermoid carcinoma cells, rat PC-12 pheochromocytoma cells, or mouse NIH3T3 fibroblasts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DSTN(11034)
mouse ... Dstn(56431)
rat ... LOC296197(296197)

General description

Destrin/ADF and cofilin are widely distributed in tissues of eukaryotes and both contain a nuclear localization sequence. It is found in various epithelial and endothelial cells but is practically absent from adult mouse heart and skeletal muscle cells. This gene is located on human chromosome 20p12.
Destrin/ADF is usually found in regions containing dynamic actin pools such as the leading edge of migrating cells and neuronal growth cones and may also colocalize in cell nuclei. It is also present in ‘Hirano bodies′ in certain brain neurons of dementia patients.

Immunogen

synthetic peptide corresponding to amino acids 137-150 of human destrin/ADF.

Application

Anti-Destrin/ADF antibody produced in rabbit is suitable for immunoblotting at a working dilution of 1:600 using whole extracts of human A-431 epidermoid carcinoma, rat PC-12 pheochromocytoma, and mouse NIH/3T3 fibroblasts, and a chemiluminescent detection reagent.

Biochem/physiol Actions

Destrin/ADF (Actin Depolymerizing Factor) is a small phosphoinositide-sensitive actin-binding protein capable of depolymerizing actin-filaments in vitro. It binds stoichiometrically to monomeric G-actin and to actin protomers in filaments in a pH-dependent, Ca2+ independent manner. It intercalates between longitudinally associated actin monomers within the filament and distorts its helical twist. It is important for many cellular processes involving actin remodeling such as motility at the leading edge of cells, polarized cell growth, endocytosis, phagocytosis, cellular activation, and cytokinesis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association study of antiphospholipid antibodies
Kamboh MI, et al.
Autoimmune diseases, 2013 (2013)
The ADF/cofilin family: actin-remodeling proteins
Maciver SK and Hussey PJ
Genome Biology, 3(5), reviews3007-reviews3001 (2002)
W Rozek et al.
Polish journal of veterinary sciences, 16(4), 663-669 (2013-01-01)
Changes in the level of cellular proteins in cells inoculated with equine influenza virus H7N7 and H3N8 were studied with microarray technique. H3N8 induced pro-apoptotic proteins while H7N7 induced both pro- as well as anti-apoptotic factors. The higher level of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service