Skip to Content
Merck
All Photos(1)

Key Documents

D7385

Sigma-Aldrich

Demecolcine

≥98% (HPLC)

Synonym(s):

N-Deacetyl-N-methylcolchicine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO5
CAS Number:
Molecular Weight:
371.43
Beilstein:
2822892
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥98% (HPLC)

form

powder

mp

184 °C

solubility

chloroform: 10 mg/mL
ethanol: soluble

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CN[C@H]1CCc2cc(OC)c(OC)c(OC)c2C3=CC=C(OC)C(=O)C=C13

InChI

1S/C21H25NO5/c1-22-15-8-6-12-10-18(25-3)20(26-4)21(27-5)19(12)13-7-9-17(24-2)16(23)11-14(13)15/h7,9-11,15,22H,6,8H2,1-5H3/t15-/m0/s1

InChI key

NNJPGOLRFBJNIW-HNNXBMFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Demecolcine has been used as a microtubule depolymerizing agent. It has also been used to inhibit mitotic division.

Biochem/physiol Actions

Often in karyotyping and cell cycle research it is desirable to increase the yield of mitotic cells in a particular phase of the cell cycle. This can be achieved in a variety of ways with the most popular being the use of a cell cycle synchronizing agent such as demecolcine. Demecolcine will arrest cells in metaphase with no remarkable effect on the biochemical events in mitotic cells or in synchronized G1 and S phase cells. White blood cells are often treated with demecolcine to arrest cells in metaphase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flow cytometric discrimination of mitotic nuclei by right-angle light scatter.
Zucker RM, et al.
Cytometry, 9, 226-231 (1988)
The Drosophila microtubule-associated protein mars stabilizes mitotic spindles by crosslinking microtubules through its N-terminal region.
Zhang G, et al.
PLoS ONE, 8, e60596-e60596 (2013)
Mathew J Thayer
BioEssays : news and reviews in molecular, cellular and developmental biology, 34(9), 760-770 (2012-06-19)
Recent studies indicate that mammalian chromosomes contain discrete cis-acting loci that control replication timing, mitotic condensation, and stability of entire chromosomes. Disruption of the large non-coding RNA gene ASAR6 results in late replication, an under-condensed appearance during mitosis, and structural
Hua Shao et al.
The Journal of cell biology, 201(2), 191-200 (2013-04-10)
The spindle assembly checkpoint (SAC) functions as a surveillance mechanism to detect chromosome misalignment and to delay anaphase until the errors are corrected. The SAC is thought to control mitosis and meiosis, including meiosis in mammalian eggs. However, it remains
Nuno Costa-Borges et al.
Theriogenology, 75(3), 527-541 (2010-11-16)
Treatment of pre-activated oocytes with demecolcine (DEM) has been shown to induce the extrusion of all oocyte chromosomes within the second polar body (PB2). However, induced enucleation (IE) rates are generally low and the competence of these cytoplasts to support

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service