Skip to Content
Merck

D7250

2′-Deoxycytidine 5′-diphosphate sodium salt

≥96%, powder

Synonym(s):

dCDP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H15N3O10P2 · xNa+
CAS Number:
Molecular Weight:
387.18 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
MDL number:
Assay:
≥96%
Form:
powder
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2′-Deoxycytidine 5′-diphosphate sodium salt, ≥96%

Quality Level

assay

≥96%

form

powder

storage temp.

−20°C

SMILES string

O.[Na].NC1=NC(=O)N(C=C1)C2CC(O)C(COP(O)(=O)OP(O)(O)=O)O2

InChI

1S/C9H15N3O10P2.Na.H2O.H/c10-7-1-2-12(9(14)11-7)8-3-5(13)6(21-8)4-20-24(18,19)22-23(15,16)17;;;/h1-2,5-6,8,13H,3-4H2,(H,18,19)(H2,10,11,14)(H2,15,16,17);;1H2;

InChI key

MNOALMYIRLFAQW-UHFFFAOYSA-N

Application

2′-Deoxycytidine 5′-diphosphate (dCDP) has been used as a substrate for nucleotide diphosphate kinase (NDPK) to produce 2′-deoxycytidine 5′-triphosphate (dCTP) in support of DNA biosynthesis and reverse transcription.


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



High glucose triggers nucleotide imbalance through O-GlcNAcylation of key enzymes and induces KRAS mutation in pancreatic cells
Hu CM, et al.
Cell Metabolism, 29(6), 1334-1349 (2019)
Argirios Siskos et al.
Journal of plant physiology, 167(9), 675-682 (2010-03-02)
Two isoforms of NDPKs (diphosphonucleoside kinases: E.C. 2.7.4.6.) named S-NDPK-A and S-NDPK-B were separated and purified from shoots of Alyssum murale (19th day of growth), a nickel accumulator plant, by a four-step procedure involving ammonium sulphate precipitation and DEAE-sepharose and
Hüseyin Besir et al.
FEBS letters, 579(29), 6595-6600 (2005-11-19)
Nucleoside diphosphate kinase from the halophilic archaeon Halobacterium salinarum was crystallized in a free state and a substrate-bound form with CDP. The structures were solved to a resolution of 2.35 and 2.2A, respectively. Crystals with the apo-form were obtained with



Global Trade Item Number

SKUGTIN
D7250-25MG04061833006979
D7250-5MG04061833006986