Skip to Content
Merck
All Photos(1)

Documents

D6442

Sigma-Aldrich

JumpStart Taq ReadyMix for High Throughput Quantitative PCR

Ready-to-use 2x mix for qPCR with ROX

Synonym(s):

Taq polymerase mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

usage

sufficient for 20 reactions
sufficient for 2000 reactions
sufficient for 400 reactions

feature

dNTPs included
hotstart

concentration

2.5 units/reaction (50 μL reaction volume)

technique(s)

qPCR: suitable

color

colorless

input

purified DNA

application(s)

agriculture

compatibility

for use with ABI 5700
for use with ABI 7000
for use with ABI 7300
for use with ABI 7500 Fast
for use with ABI 7500
for use with ABI 7700
for use with ABI 7900 Fast
for use with ABI 7900 HT
for use with ABI 7900
for use with ABI StepOne
for use with ABI StepOnePlus
for use with ABI ViiA 7
for use with Bio-Rad CFX384
for use with Bio-Rad CFX96
for use with Bio-Rad MJ Chromo4
for use with Bio-Rad MJ Opticon 2
for use with Bio-Rad MJ Opticon Cepheid SmartCycler
for use with Bio-Rad MJ Opticon
for use with Bio-Rad MiniOpticon
for use with Eppendorf® Mastercycler ep realplex2 s
for use with Eppendorf® Mastercycler ep realplex
for use with Illumina Eco qPCR
for use with Qiagen Corbett Rotor-Gene 3000
for use with Qiagen Corbett Rotor-Gene 6000
for use with Qiagen Corbett Rotor-Gene Q
for use with Roche LightCycler 480
for use with Strategene Mx3000P
for use with Strategene Mx3005P
for use with Strategene Mx4000

detection method

probe-based

shipped in

wet ice

storage temp.

−20°C

General description

JumpStart Taq ReadyMix For High Throughput Quantitative PCR combines the performance enhancements of JumpStart Taq Antibody for hot start PCR with the convenience of an easy-to-use reaction mixture that incorporates the internal reference dye for ABI and other real time instrument applications. This 2× concentrate master mix contains JumpStart Taq DNA polymerase, 99% pure deoxynucleotides and reaction buffer. Simply add an equal volume of the 2× ReadyMix to a 2× mixture DNA template, primers and fluorescent probe.
To prepare a typical qPCR reaction, mix 25 μL of JumpStart ReadyMix Taq, fluorescent probe, desired amount of magnesium chloride (above 1.5 mM final concentration), template DNA, and primers in a final volume of 50 μL. Reaction volumes can be scaled down, if desired.

Application

JumpStart Taq ReadyMix for High Throughput Quantitative PCR has been used:
  • For quantitative polymerase chain reaction (qPCR) of cDNA template
  • As a component of PCR reaction mix for amplification of DNA extracted from peripheral blood leucocytes
  • For qPCR amplifications using SYBR
  • For multiplex PCR
  • For reduction of primer dimers

Features and Benefits

  • JumpStart Taq DNA polymerase prevents amplification of non-specific products, resulting in increased efficiency and higher target yield
  • When performing large number of PCR reactions, JumpStart Taq ReadyMix reduces preparation time and the risk of contamination from multiple pipetting steps
  • This master mix allows consistent batch-to-batch and reaction-to-reaction performance
  • Internal Reference Dye is provided for reaction normalization

Packaging

Default reaction volume is 50 μL

20RXN is packaged as 1 X 500 μL
400RXN is packaged as 1 X 10 mL
2000RXN is packaged as 1 X 50 mL

Quantity

Use 2.5 units per reaction.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
JumpStart is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Ox. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edouard Jurkevitch
Current protocols in microbiology, Chapter 7, Unit7B-Unit7B (2012-08-10)
Bdellovibrio and like organisms (BALOs) are obligate predators of Gram-negative bacteria. BALOs are isolated as plaques growing at the expense of their prey and are cultivated as two-member cultures. The growth cycle is composed of an extracellular attack phase and
Adrien Chabot et al.
Genetics, 176(4), 2069-2076 (2007-06-15)
Most phenotypic differences between human and chimpanzee are likely to result from differences in gene regulation, rather than changes to protein-coding regions. To date, however, only a handful of human-chimpanzee nucleotide differences leading to changes in gene regulation have been
Evrim Komurcu-Bayrak et al.
Clinica chimica acta; international journal of clinical chemistry, 383(1-2), 110-115 (2007-06-09)
We evaluated the relationship of the lipoprotein lipase (LPL) S447X variant with serum lipid levels and the metabolic syndrome (MS) in the Turkish Adult Risk Factor (TARF) study. This is the first study examining this LPL variant in the Turkish
Irene B Hanning et al.
Methods in molecular biology (Clifton, N.J.), 733, 159-170 (2011-03-25)
Next-generation sequencing (NGS) is a powerful tool that can be utilized to profile and compare microbial populations. By amplifying a target gene present in all bacteria and subsequently sequencing amplicons, the bacteria genera present in the populations can be identified
Malgorzata Gozdecka et al.
Cell reports, 9(4), 1361-1374 (2014-12-03)
JNK and p38 phosphorylate a diverse set of substrates and, consequently, can act in a context-dependent manner to either promote or inhibit tumor growth. Elucidating the functions of specific substrates of JNK and p38 is therefore critical for our understanding

Articles

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

See All

Related Content

RT-qPCR detects specific targets with applications in gene expression and pathogen detection.

RT-qPCR detects specific targets with applications in gene expression and pathogen detection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service