Skip to Content
Merck
All Photos(1)

Documents

D5773

Sigma-Aldrich

Dulbecco′s Phosphate Buffered Saline

Without calcium chloride, powder, suitable for cell culture

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
eCl@ss:
32129211
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Use this formulation of DPBS when tissue or cell dissociation or release is the objective. Also, suitable for studies that involve calcium metabolism.

Quantity

Formulated to contain 9.6 grams of powder per liter of medium.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Christine MacKay et al.
PloS one, 9(5), e96573-e96573 (2014-05-08)
Stressor experiences during the juvenile period may increase vulnerability to anxiety and depressive-like symptoms in adulthood. Stressors may also promote palatable feeding, possibly reflecting a form of self-medication. The current study investigated the short- and long-term consequences of a stressor
Matheus Pereira Dos Santos et al.
Zygote (Cambridge, England), 24(6), 795-807 (2016-11-03)
The aim of this study was to describe the morphology of gametes, post-fertilization events and subsequent temperature effects on the early developmental stages of the neotropical species Astyanax altiparanae. The sperm of this species presents a typical morphology of teleost
Enrique Valera et al.
Clinical biochemistry, 49(1-2), 121-126 (2015-09-15)
We report the development of an optical immunosensor for the detection of monocyte chemotactic protein 1 (MCP-1) in serum samples. MCP-1 is a cytokine that is an emerging biomarker for several diseases/disorders, including ischemic cardiomyopathy, fibromyalgia, and some cancers. The
Regiane Cristina da Silva et al.
Zygote (Cambridge, England), 25(5), 637-651 (2017-09-21)
Primordial germ cell (PGC) transplant is a promising tool in aquaculture; however, successful use of this technique requires in depth knowledge of the early stages of embryo and larval development. The aim of this study was to analyse the effect
Eliza Fatima Ali et al.
Frontiers in behavioral neuroscience, 12, 216-216 (2018-10-05)
The juvenile period is marked by a reorganization and growth of important brain regions including structures associating with reward seeking behaviors such as the nucleus accumbens (NA) and prefrontal cortex (PFC). These changes are impacted by stressors during the juvenile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service