Skip to Content
Merck

Skip To

D4947

DMEM - low glucose

With ʟ-glutamine and sodium bicarbonate, without phenol red and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, L-glutamine and sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

glucose: low
sodium pyruvate: no
phenol red: no
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

General description

This modification has been shown to improve the stability of the medium.

Application

Dulbecco′s Modified Eagle′s Medium - low glucose has been used to grow human cell line EA.hy926.

Other Notes

Investigators who work with stem cells or cell clones frequently prefer media without phenol red.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0415D5921D9443
sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

components

glucose: low
sodium pyruvate: no
phenol red: no
L-glutamine: yes

components

L-glutamine: no
NaHCO3: yes
HEPES: no
glucose: low
phenol red: yes
sodium pyruvate: yes

components

phenol red: no
sodium pyruvate: no
glucose: low
HEPES: no
L-glutamine: no
NaHCO3: yes

components

NaHCO3: 3.7 g/L
sodium pyruvate: no
glucose: 1.0 g/L
L-glutamine: 0.584 g/L
phenol red: no
HEPES: no

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters