Skip to Content
Merck
All Photos(2)

Key Documents

D3159

Sigma-Aldrich

Deoxyribonucleic acid from herring sperm

not suitable for substrate for typical DNase assays (“crude oligonucleotides", <50 bp), degraded

Synonym(s):

Deoxyribonucleic acid, partially degraded from herring sperm, hsDNA

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
NACRES:
NA.51

form

powder

suitability

not suitable for substrate for typical DNase assays (“crude oligonucleotides", <50 bp)

storage temp.

2-8°C

General description

Herring Sperm DNA, also known as hsDNA is a natural source of DNA designed for DNA binding agent studies. (1) The detection limits of hsDNA are around 5.0ng and linear ranges of light scattering intensity (I(LS)) is 0.05-2.5 mµg.

Application

Herring sperm DNA (hsDNA) is a natural DNA used in studies of DNA binding anticancer agents and DNA binding agents that modulate DNA structure and function. Herring sperm DNA is also used in physicochemical studies of DNA behavior in solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanyan Niu et al.
Journal of chromatography. A, 1463, 102-109 (2016-08-16)
Fufang Banbianlian Injection (FBI) has been widely used as an anti-inflammatory and anti-tumor prescription. To understand the relationships between its bioactive ingredients and pharmacological efficacies, our previous study has been successfully identified some DNA-binding compounds in FBI using an established
Yan Zhang et al.
Nucleosides, nucleotides & nucleic acids, 30(1), 49-62 (2011-01-25)
The Mg(II) complex of Schiff base (K[HL]) derived from vanillin and L-tryptophan could bind with herring sperm DNA. The binding behaviors between them in physiological pH environment (pH 7.40) have been studied by spectroscopy, cyclic voltammetry and viscosity methods. Binding
Po Wang et al.
Biosensors & bioelectronics, 26(7), 3339-3345 (2011-02-08)
A rapid, convenient and accurate method for the simultaneous detection of guanine (G), adenine (A), thymine (T) and cytosine (C) was developed at a multiwalled carbon nanotube (MWCNT)/choline (Ch) monolayer-modified glassy carbon electrode (GCE). X-ray photoelectron spectroscopy data demonstrated that
Synthesis, spectroscopic, crystal structure and DNA binding of Ru(II) complexes with 2-hydroxy-benzoic acid [1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)-ethylidene]-hydrazide.
Chitrapriya N, Sathiya Kamatchi T, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 81, 128-134 (2011)
Mechanistic pathway for controlled extraction of guest molecule bound to herring sperm DNA using α-cyclodextrin.
Jaffer SS, Ghosh P, Purkayastha P.
Spectrochimica Acta Part A: Molecular Spectroscopy, 78, 1587-1591 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service