Skip to Content
Merck
All Photos(2)

Key Documents

D2396

Sigma-Aldrich

Dimenhydrinate

Synonym(s):

N-(2-Diphenylmethoxyethyl)-N,N-dimethylammonium 8-chlorotheophyllinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · C7H7ClN4O2
CAS Number:
Molecular Weight:
469.96
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

SMILES string

CN1C(=O)N(C)c2nc(Cl)[nH]c2C1=O.CN(C)CCOC(c3ccccc3)c4ccccc4

InChI

1S/C17H21NO.C7H7ClN4O2/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h3-12,17H,13-14H2,1-2H3;1-2H3,(H,9,10)

InChI key

NFLLKCVHYJRNRH-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Dimenhydrinate is a salt of antihistamine drugs, such as diphenhydramine and 8-chlorotheophylline. It has antiemetic properties. Dimenhydrinate is used to treat nausea and vomiting during pregnancy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerard Nobel et al.
Brain research bulletin, 82(3-4), 193-200 (2010-04-17)
Motion sickness (MS) exaggerates body cooling during cold-water immersion. The aim of the present study was to investigate whether such MS-induced predisposition to hypothermia is influenced by two anti-MS drugs: the histamine-receptor blocker dimenhydrinate (DMH) and the muscarine-receptor blocker scopolamine
Serge Gouin et al.
Pediatrics, 129(6), 1050-1055 (2012-05-16)
To evaluate the efficacy and safety of oral dimenhydrinate in the treatment of acute gastroenteritis. This was a randomized, double-blind, placebo-controlled trial conducted in the emergency department of a pediatric university-affiliated center. Children 1 to 12 years old who presented
Xiang Li et al.
The Journal of chemical physics, 133(14), 144304-144304 (2010-10-19)
The hydrated nucleoside anions, uridine(-)(H(2)O)(n=0-2), cytidine(-)(H(2)O)(n=0-2), and thymidine(-)(H(2)O)(n=0,1), have been prepared in beams and studied by anion photoelectron spectroscopy in order to investigate the effects of a microhydrated environment on parent nucleoside anions. Vertical detachment energies (VDEs) were measured for
Tawnya G Flick et al.
Analytical chemistry, 82(4), 1179-1182 (2010-01-23)
Accurate and rapid quantitation is advantageous to identify counterfeit and substandard pharmaceutical drugs. A standard-free electrospray ionization mass spectrometry method is used to directly determine the dosage in the prescription and over-the-counter drugs Tamiflu, Sudafed, and Dramamine. A tablet of
Drugs During Pregnancy and Lactation: Handbook of Prescription Drugs and Comparative Risk Assessment (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service