Skip to Content
Merck
All Photos(1)

Key Documents

D0815

Sigma-Aldrich

2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide

≥99% purity (HPLC), solid

Synonym(s):

DASPEI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21IN2
CAS Number:
Molecular Weight:
380.27
Beilstein:
3783243
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide, ≥99% (HPLC), solid

Quality Level

Assay

≥99% (HPLC)

form

solid

color

red

mp

267 °C (dec.) (lit.)

solubility

DMSO: 0.5 mg/mL
methanol: soluble

εmax

36.0-41.0 at 457-463 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[I-].CC[n+]1ccccc1\C=C\c2ccc(cc2)N(C)C

InChI

1S/C17H21N2.HI/c1-4-19-14-6-5-7-17(19)13-10-15-8-11-16(12-9-15)18(2)3;/h5-14H,4H2,1-3H3;1H/q+1;/p-1

InChI key

AMAXNNVXIBDEMV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide (DASPEI) has been used for the labelling of hair cells in larval zebrafish.

Biochem/physiol Actions

2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide (DASPEI) accumulates in the mitochondria and has a large fluorescence Stokes shift. It is also used for the relative measurements of mitochondrial membrane potential.
Suitable as a mitochondrial stain in live cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screen of FDA-approved drug library reveals compounds that protect hair cells from aminoglycosides and cisplatin.
Vlasits AL, et al.
Hearing Research, 294, 153-165 (2012)
Ru Zhang et al.
Biomolecules, 10(10) (2020-10-22)
Cochlear hair cells in human beings cannot regenerate after loss; however, those in fish and other lower species can. Recently, the role of inflammation in hair cell regeneration has been attracting the attention of scientists. In the present study, we
Development of a no-wash assay for mitochondrial membrane potential using the styryl dye DASPEI.
Jensen KH and Rekling JC
Journal of Biomolecular Screening, 15, 1071-1081 (2010)
Marina Yoffe et al.
Journal of experimental zoology. Part B, Molecular and developmental evolution, 334(7-8), 511-517 (2020-05-22)
The lateral line is the primary modality fish use to create a hydrodynamic image of their environment. These images contribute to a variety of behaviors, from rheotaxis to escape responses. Here we discern the contributions of visual and lateral line
E G Sedgwick et al.
Biochimica et biophysica acta, 1099(1), 51-56 (1992-01-30)
The mechanism of uptake of the fluorescent dye 2-(4-dimethylaminostyryl)-1-ethylpyridinium cation (DMP+) into cells and vesicles of the acrA strain AS-1 of Escherichia coli was examined. Uptake was energized by substrate oxidation and discharged by uncouplers. Uptake was enhanced by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service