Skip to Content
Merck
All Photos(3)

Key Documents

D0776

Sigma-Aldrich

2′-Deoxycytidine hydrochloride

BioReagent, suitable for cell culture

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)cytosine hydrochloride, Cytosine deoxyriboside hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O4 · HCl
CAS Number:
Molecular Weight:
263.68
Beilstein:
3576124
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

product line

BioReagent

Assay

≥99% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

water: 50 mg/mL, clear, colorless

SMILES string

Cl[H].NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](CO)O2

InChI

1S/C9H13N3O4.ClH/c10-7-1-2-12(9(15)11-7)8-3-5(14)6(4-13)16-8;/h1-2,5-6,8,13-14H,3-4H2,(H2,10,11,15);1H/t5-,6+,8+;/m0./s1

InChI key

LTKCXZGFJFAPLY-OERIEOFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′-Deoxycytidine hydrochloride has been used:
  • as a deoxyribonucleoside component in the basal medium to study its effects on Bacillus mojavensis′s anaerobic growth
  • in the infectivity assay of monocyte-derived macrophages by human immunodeficiency virus 1 (HIV-1)
  • in nucleoside supplementation for the synchronization of human retinal pigment epithelial-1 (RPE-1) cells in G1 phase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuxia Li et al.
American journal of cancer research, 10(3), 908-924 (2020-04-09)
Epithelial ovarian cancer is characterized by universal TP53 mutations, which result in G1/S checkpoint deficiencies. Therefore, it is hypothesized that the abrogation of the G2/M checkpoint with Wee1 inhibitor might preferentially sensitize TP53-defective ovarian cancer cells. Given the extremely high
Md Maminur Rahman et al.
The Journal of biological chemistry, 295(51), 17460-17475 (2021-01-18)
Homologous recombination (HR) repairs DNA double-strand breaks using intact homologous sequences as template DNA. Broken DNA and intact homologous sequences form joint molecules (JMs), including Holliday junctions (HJs), as HR intermediates. HJs are resolved to form crossover and noncrossover products.
Aiman A Haqqani et al.
Virology journal, 12, 184-184 (2015-11-13)
Template switching between two distinct HIV-1 RNA genomes during reverse transcription gives rise to recombinant viruses that greatly expand the genetic diversity of HIV-1 and have adverse implications for drug resistance, immune escape, and vaccine design. Virions with two distinct
Kyosuke Nakamura et al.
Molecular cell, 81(5), 1084-1099 (2021-01-16)
Cells have evolved an elaborate DNA repair network to ensure complete and accurate DNA replication. Defects in these repair machineries can fuel genome instability and drive carcinogenesis while creating vulnerabilities that may be exploited in therapy. Here, we use nascent
null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service