Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.51
UNSPSC Code:
41106609
Concentration:
20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)
Pricing and availability is not currently available.
form
liquid
Quality Segment
packaging
vial of 50 μL
concentration
20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)
application(s)
CRISPR
shipped in
dry ice
storage temp.
−20°C
General description
Universal negative control CRISPRs have been designed not to recognize any sequence in the human, mouse or rat genome. A single vector format is provided which includes the Cas9 expression cassette and gRNA sequence. This vector includes GFP which is co-expressed from the same mRNA as the Cas9 protein via a 2A peptide linkage and enables for tracking of transfection efficiency or enrichment in cell populations via FACS.
Application
Functional Genomics/Target Validation
- Creation of gene knockouts in cell lines
- Creation of knock-in cell lines with promoters, fusion tags or reporters integrated into endogenous genes
Biochem/physiol Actions
CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.
Features and Benefits
Allows for expression of Cas9 and GFP without specific targeting of the CRISPR/Cas9 system.
Physical form
Sigma U6-gRNA/CMV-Cas9-GFP plasmid expressing a non-targeting guide sequence supplied at a concentration of 20ng/ul in 50ul.
Preparation Note
Sigma CRISPR plasmid products are delivered as mini-prep aliquots,
which may not be suitable for transfection into particular cell types. For best results, we advise maxi-prepping
plasmids using endotoxin-free DNA purification kits prior to transfection.
which may not be suitable for transfection into particular cell types. For best results, we advise maxi-prepping
plasmids using endotoxin-free DNA purification kits prior to transfection.
Other Notes
1 vial containing 1ug of U6-gRNA/CMV-Cas9-GFP plasmid expressing a non-targeting guide sequence.
Typical transfection concentrations used in literature are in the ranges of 2.0 to 8.0ug of the single vector expressing the guideRNA and Cas9 . (All dosages above assume 0.5 to 1 million cells nucleofected)
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| concentration 20 ng/μL in TE buffer; DNA (1μg of plasmid DNA) | concentration 20 ng/μL in TE buffer; DNA (1μg of plasmid DNA) | concentration 20 ng/μL in TE buffer; DNA (1μg of plasmid DNA) | concentration 20 ng/μL in TE buffer; DNA (1μg of plasmid DNA) |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| packaging vial of 50 μL | packaging vial of 50 μL | packaging vial of 50 μL | packaging vial of 50 μL |
| application(s) CRISPR | application(s) CRISPR | application(s) CRISPR | application(s) CRISPR |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



