Skip to Content
Merck
All Photos(1)

Key Documents

CLS4863

Corning® universal fit pipet tips

1-200 μL universal fit racked pipet tips, natural, non-sterile, 10 racks/case, 960 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation
96 Tips/Rack

packaging

rack of 96

manufacturer/tradename

Corning 4863

volume range

1-200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Universal Fit Pipet Tips

Corning Universal Fit ploypropylene pipet tips are designed to provide a reliable fit with all major brand pipettors and have a bevelled orifice to help ensure fluid delivery. All racked tips are certified RNase/DNase free and nonpyrogenic.

  • Reliable fit with all major brand pipettors
  • Bevelled orifice to help ensure fluid delivery
  • Certified RNase/DNase free and nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service