Skip to Content
Merck
All Photos(1)

Key Documents

CLS480133

Corning® LSE dual block, for Corning LSE dual block digital dry baths

dual, block, 384 well PCR plates, 1/cs

Synonym(s):

equipment replacement accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NB.15

feature

Holds 2 x 96-well plates

packaging

case of 1
pack of 1

manufacturer/tradename

Corning 480133

technique(s)

PCR: suitable

suitability

suitable for (use with Corning LSE digital dry bath heater, dual block only)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service