Skip to Content
Merck

CLS4591

Corning® 96 well plates

round bottom black, ULA, lid, sterile

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41121800
Material:
round bottom black
Size:
96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

round bottom black

sterility

sterile

feature

lid, skirt, ULA

packaging

pack of 1 ea, case of 25 ea

manufacturer/tradename

Corning 4591

size

96 wells

well volume

360 μL

wells

96 wells

working volume

75-200 μL

suitability

suitable for (cell cuture)

binding type

(ultra-low attachment)

General description

  • Corning® 96 well plates are general assay plates, ideal for solution-based assays.
  • Ultra-low attachment surface has a neutral, hydrophilic hydrogel coating which reduces binding of attachment proteins, minimizing cell attachment and spreading.
  • The surface is noncytotoxic, biologically inert, and nondegradable.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Designed to reduce well-to-well cross-talk.
  • Conforms to standard microplate footprint and dimensions
  • Sterilized by gamma radiation and nonpyrogenic.

Application

These plates are general assay plates, ideal for solution-based assays (i.e. hemagglutination or precipitation assays), serial dilutions and general storage applications.

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® 96 well plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library