CLS4571
Corning® 1536 well plates, cyclic olefin co-polymer
white, tissue-culture treated, w/o lid, sterile
Synonym(s):
1536 multiwell plate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.13
material:
white
white bottom
white bottom
well volume:
12.5 μL
wells:
1536 wells
working volume:
0-8 μL
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid: no
skirt
skirt
Recommended Products
material
white
white bottom
sterility
sterile
feature
lid: no
skirt
packaging
pack of 10 ea
case of 50 ea
manufacturer/tradename
Corning 4571
well volume
12.5 μL
wells
1536 wells
working volume
0-8 μL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
1536 well cyclic olefin copolymer microplates are perfectly suited for high-throughput screening (HTS). A variety of surface treatments enable both cell-based and biochemical assays. The ultraclear, 127 μm film bottom thickness is ideal for cell-based assays that require advanced optical analysis. The COC microplates are mechanically stable and inherently resistant to a broad range of solvents including DMSO and alcohols, allowing for compound storage as well.
Key Features
Key Features
- Cyclic Olefin Copolymer (COC) material
- Flat top for improved sealing
- 1536 Well Hi Base solid color microplates (black or white)
- Variety of treatment options
- tissue culture treated
- Corning CellBIND® Surface
- Nonbinding Surface (NBS)
- Not treated
- Bar coded (one label on both long sides)
- Custom bar codes available for compatibility with the Kalypsys System
- Low autofluorescence
- Broad chemical resistance including DMSO and alcohol, which allows for assay step reduction
- High mechanical stability
- Optimized for flatness and uniformity
- Low birefringence
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service