Skip to Content
Merck
All Photos(1)

Key Documents

CLS4511

Corning® 384 well microplate, low volume

black polystyrene, round bottom, non-sterile, lid: no, non-treated, pack of 10

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
black bottom
black polystyrene
round bottom
well volume:
35 μL
wells:
384 wells , low volume
working volume:
2-20 μL
sterility:
non-sterile
binding type:
non-treated surface
feature:
lid: no
skirt
non-treated

material

black bottom
black polystyrene
round bottom

sterility

non-sterile

feature

lid: no
skirt
non-treated

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 4511

well volume

35 μL

wells

384 wells , low volume

working volume

2-20 μL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Corning®384 well microplate, low volume is suitable for immobilizing large molecules with hydrophobic surface-interaction regions, such as antibodies.

Features and Benefits

  • Square well top enables easier robotic pipetting
  • Light cone-shaped conical well for better efficiency
  • Rounded bottom to minimize trapped air and increase the Z factor
  • In fluorescent assays, black walls minimize autofluorescence and crosstalk between well
  • Biomolecules are bound by passive interactions on an untreated polystyrene surface due to its hydrophobic nature

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service