CLS432055
Corning® CoolRack®
XT PCR384
Synonym(s):
Staining, Staining Accessories
About This Item
Recommended Products
material
foamed PE
feature
wells 384
Holds 1 x 384 well plate
packaging
pkg of 1
manufacturer/tradename
Corning 432055
technique(s)
PCR: suitable
cryopreservation: suitable
L × W × H
12.7 cm × 8.6 cm × 1.9 cm
size
4.5 mm , column spacing
4.5 mm , row spacing
well depth
7.6 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service