Skip to Content
Merck
All Photos(1)

Key Documents

CLS3770

Corning® CellBIND® 384 well microplate

CellBIND® surface, black polystyrene, flat bottom, sterile, lid, pack of 20

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
black plate (clear bottom)
black polystyrene
clear bottom
flat bottom
well volume:
112 μL
wells:
384 wells
working volume:
20-80 μL

material

black plate (clear bottom)
black polystyrene
clear bottom
flat bottom

sterility

sterile

feature

lid
skirt
plate format: low flange, with lid

packaging

case of 100
pack of 20

manufacturer/tradename

Corning 3770

well volume

112 μL

wells

384 wells

working volume

20-80 μL

color

black

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® CellBIND® 384 well microplate is low flange, black plate with a clear bottom and a flat polystyrene surface. The plate is designed to minimize autofluorescence and optimize signal to background ratios, resulting in accurate and reliable assay results. Additionally, the enhanced flatness of the plates ensures low variations within and between plates, making them compatible with automated liquid dispensers for efficient and precise sample handling. The inclusion of a lid and skirt provides added protection and maintains sterility throughout the experiment. Each plate is provided in a sterile condition.

Application

Corning® CellBIND® 384 well microplate has been used for the bottom and top reading instruments and to measure adipocyte differentiation and cell proliferation.

Features and Benefits

  • Exhibit minimal background fluorescence
  • Reduced light scattering thereby minimizing crosstalk between wells. 20% higher signal to background ratio
  • Clear and flat well bottom, allowing direct microscopic viewing of cells and samples without compromising clarity.
  • Compatible with Top and Bottom Reading Instruments
  • The microplates are treated with Corning CellBIND® surface treatment, promoting optimal cell attachment for reliable cell-based assays.
  • Gamma Radiation Sterilization and Nonpyrogenic
  • Wells are assigned a unique alphanumeric code, enabling easy well identification

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Merrick et al.
Science (New York, N.Y.), 364(6438) (2019-04-27)
Metabolic health depends on the capacity of adipose tissue progenitor cells to undergo de novo adipogenesis. The cellular hierarchy and mechanisms governing adipocyte progenitor differentiation are incompletely understood. Through single-cell RNA sequence analyses, we show that the lineage hierarchy of
Anthony R Angueira et al.
Nature metabolism, 3(4), 469-484 (2021-04-14)
Brown adipose tissue can expend large amounts of energy, and therefore increasing its size or activity is a promising therapeutic approach to combat metabolic disease. In humans, major deposits of brown fat cells are found intimately associated with large blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service