Skip to Content
Merck
All Photos(1)

Key Documents

CLS3569

Corning® disposable spinner flask, with aseptic, 1 L

Synonym(s):

Corning spinner flask, Disposable spinner flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

high-density polyethylene cap
polystyrene
screw top cap

sterility

sterile; γ-irradiated

feature

cap
closure type Screw cap

packaging

case of 6

manufacturer/tradename

Corning 3569

availability

available only in South Korea

capacity

1000 mL

working volume

300-1000 mL

fitting

cap for GL 45
GL 45 neck

Features and Benefits

  • Each system is packaged in a separate, double bag
  • Flask body is made up of polystyrene, caps from high-density polyethylene, and polypropylene is used to make the paddle and shaft
  • Side arm of a preassembled flask has one connector attached with a flat cap on an opposite side arm
  • Without having to adjust the height of the paddle, the extended paddle design ensures constant liquid motion from top to bottom
  • Dead spots are removed by the paddle′s bottom contours, which hug the flask′s shape
  • Two large neck openings allow for easy pouring
  • To ensure better mixing and maximize cell production vertical side baffles break up laminar flow
  • Gamma-irradiated sterilized with sterility assurance level (SAL) of 10-6 and meets the bacteriostasis, fungistasis, and sterility standards of USP XXIV:71
  • A 25mm-diameter vent filter with a polyvinylidene fluoride (PVDF) membrane and 0.2-micron pores and a barbed fitting completes the venting port
  • Only for single use, flasks cannot be alcohol-resterilized or autoclaved, materials will deteriorate in any attempt to do so
  • Aseptic transfer and vented caps are available optionally
  • A rigid polypropylene dip tube that is angled to reach the flask′s bottom is attached to the liquid transfer port′s internal end
  • Compatible with corning microcarriers in both suspension and adherent cell culture applications
  • A heat-sealable and chemically resistant tubing ends in a male luer lock (MLL) connector closed with a female luer sealing cap on the external side

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Natascha Verhagen et al.
FEBS open bio, 10(12), 2791-2804 (2020-11-01)
A major goal for process and cell engineering in the biopharmaceutical industry is enhancing production through increasing volumetric and cell-specific productivities (CSP). Here, we present 5'-deoxy-5'-(methylthio)adenosine (MTA), the degradation product of S-(5'-adenosyl)-L-methionine (SAM), as a highly attractive native additive which
Kerstin Schneeberger et al.
Hepatology (Baltimore, Md.), 72(1), 257-270 (2019-11-13)
The gap between patients on transplant waiting lists and available donor organs is steadily increasing. Human organoids derived from leucine-rich repeat-containing G protein-coupled receptor 5 (LGR5)-positive adult stem cells represent an exciting new cell source for liver regeneration; however, culturing
Juuso Rusanen et al.
Viruses, 13(2) (2021-01-28)
Accurate and rapid diagnostic tools are needed for management of the ongoing coronavirus disease 2019 (COVID-19) pandemic. Antibody tests enable detection of individuals past the initial phase of infection and help examine vaccine responses. The major targets of human antibody
Eliza J R Peterson et al.
Cell reports, 31(4), 107577-107577 (2020-04-30)
Mycobacterium tuberculosis (MTB) displays the remarkable ability to transition in and out of dormancy, a hallmark of the pathogen's capacity to evade the immune system and exploit susceptible individuals. Uncovering the gene regulatory programs that underlie the phenotypic shifts in
Matthias Romauch
Open biology, 10(4), 190035-190035 (2020-04-22)
Zinc-α2-glycoprotein (ZAG) is a major plasma protein whose levels increase in chronic energy-demanding diseases and thus serves as an important clinical biomarker in the diagnosis and prognosis of the development of cachexia. Current knowledge suggests that ZAG mediates progressive weight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service