Skip to Content
Merck
All Photos(1)

Key Documents

CLS2482

Corning® 1 x 8 Stripwell 96 well plates

low volume, medium binding surface, black

Synonym(s):

1 x 8 strip plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

black polystyrene plate
flat bottom clear polystyrene (96-well)
round black wells

sterility

non-sterile

feature

lid: no
skirt

packaging

plate of 12 strips

manufacturer/tradename

Corning 2482

size

96 wells

well volume

190 μL

well working volume

75-200 μL , for low volume plates volume is 50-125 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The Costar 1 × 8 strips are pre-assembled in an "egg crate" style 96 well strip holder. Strips fit in holder only one way, eliminating mis-orientation. If covers are needed, the notched cover fits the fully assembled strip holder.

Features and Benefits

  • Corning® Stripwell 96 well plates are designed for in vitro diagnostic assays such as immobilization of antibodies.
  • Non-treated medium binding hydrophobic surface binds biomolecules through passive interactions.
  • Black wells have low background fluorescence, minimal light scatter, and reduced crosstalk.

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service