Skip to Content
Merck
All Photos(2)

Key Documents

CHODHFR

SAFC

CHOZN® ZFN-Modified CHO Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.75

description

CHOZN® DHFR-/- ZFN-modified CHO cell line
dihydrofolate reductase knockout cell line

Quality Level

shipped in

dry ice

storage temp.

−196°C

General description

CHOZN cell lines were created utilizing Sigma′s proprietary CompoZr Zinc Finger Nuclease (ZFN) technology to create genetic modifications in parental CHO-K1 cells. Cells are banked in chemically defined, animal component free EX-CELL CD CHO Fusion media

Cell Line Origin

CHOZN cell lines are all subclones of the parental CHO-K1 cell line originated by Puck in 1957.

Physical form

CHOZN cell lines are provided to customers in vials containing >7.5e6 cells/mL

Legal Information

All CHOZN cell lines are sold under license from Sigma Aldrich. A copy of the research license is provided with purchase of the CHOZN cell lines.
CHOZN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MGAT1 adds N-acetylglucosamine to the Man5GlcNAc2 (Man5) structure. Goh et al. reported increased sialylation after restoring MGAT1 function in MGAT1 deficient CHO cells.

A signal peptide is a 5-30 amino acid (aa) peptide present at the N-terminus of secretory proteins.

Related Content

Learn more about cell line development and the relevant products you need for this process.

Learn more about cell line development and the relevant products you need for this process.

Learn more about cell line development and the relevant products you need for this process.

Learn more about cell line development and the relevant products you need for this process.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service