DMS 273
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 95062830, human lung (effusion), Not specified
Synonym(s):
DMS273 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human lung (effusion)
packaging
tube of 5 μg 95062830-DNA-5UG
pkg of vial of cells 95062830-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human lung, small cell carcinoma
Cell Line Description
DMS 273 was initiated in 1978 from the pleural fluid specimen from a 50 year old female patient with small cell carcinoma of the lung. The patient was in relapse after chemotherapy and radiotherapy. DMS 273 cells exhibit intercellular processes and express retinoblastoma mRNA and protein. The cell line is tumorigenic in nude mice.
Application
Human lung tumour model
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10 ,12
D13S317: 12,13
D16S539: 11
D5S818: 11,12
D7S820: 10,12
THO1: 9.3
TPOX: 10
vWA: 15
CSF1PO: 10 ,12
D13S317: 12,13
D16S539: 11
D5S818: 11,12
D7S820: 10,12
THO1: 9.3
TPOX: 10
vWA: 15
Culture Medium
Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service