Skip to Content
Merck
All Photos(1)

Documents

JIYOYE

88071302, human lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

JIYOYE, 88071302

biological source

human lymph

growth mode

Suspension

karyotype

2n = 46, diploid

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Burkitt′s lymphoma

Cell Line Description

Derived from the ascitic fluid of a male with Burkitt′s lymphoma of the liver. The cells contain an unidentified herpes-like virus and have been used in co-cultivation studies to establish lymphocytes in cultures. The cell line is also EBNA positive. Ethnicity: Black.

Application

Co-cultivation and virus studies: Herpesvirus

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 12
D16S539: 10,11
D5S818: 12
D7S820: 8,10
THO1: 7,9
TPOX: 6,8
vWA: 15,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 4-6x100,000 cells/ml. Add fresh medium every 2-3 days. Maximum cell density is achieved at 2-3x1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service