Skip to Content
Merck
All Photos(1)

Key Documents

94A1

85062503, Application: Fusion partner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Bovine Heterohybridoma

ImmunoDon

Jersey calf lymph node

Myeloma

94A1 (mouse x bovine heteromyeloma)

Reactivity

Application: Fusion partner

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse 53B3 x Bovine Jersey calf lymph node cells

Cell Line Description

The antibody is directed against sheep red blood cells. It does not react with bovine red cells. This haemocytic antibody is probably directed against the Forssman hapten. This HAT sensitive line can be used as a fusion partner for the production of heterohybridomas.

Immunogen

Sheep red blood cells

Application

Fusion partner
The isotype of the monoclonal antibody produced by this cell line is bovine IgG1.

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Related Content

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service