Skip to Content
Merck
All Photos(1)

Key Documents

HeB5

11033104, mouse liver, Epithelial

Synonym(s):

Hepa-1c1c7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HeB5, 11033104

biological source

mouse liver

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Hepa-1c1c7, genetic modification, Xenobiotic-Responsive Element (XRE)

Cell Line Description

HeB5 is a derivative of Hepa-1c1c7 (Sigma product number 95090613) cell line stably transfected with a firefly luciferase reporter construct bearing Xenobiotic-Responsive Element (XRE) driven by a TATA promoter. HeB5 can be used for detection of mouse CYP1A induction by chemicals.

Culture Medium

Alpha-MEM + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442) + 16μg/ml Blasticidin S HCL.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10, i.e. seeding at 2 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service