COV318
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 7071903, human ovary, Polygonal & fusiform
Synonym(s):
COV-318 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human ovary
packaging
tube of 5 μg 07071903-DNA-5UG
pkg of vial of cells 07071903-1VL
growth mode
Adherent
karyotype
Modal chromosome no. 74 (68-77)
morphology
Polygonal & fusiform
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human ovarian epithelial-serous carcinoma, peritoneal ascites
Cell Line Description
A human ovarian epithelial-serous carcinoma cell line established from a peritoneal ascites. Anchorage-independent growth in agar has been reported.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 14
D16S539: 9,12
D5S818: 11,13
D7S820: 9
THO1: 9.3
TPOX: 8
vWA: 15
CSF1PO: 11,13
D13S317: 14
D16S539: 9,12
D5S818: 11,13
D7S820: 9
THO1: 9.3
TPOX: 8
vWA: 15
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service