Skip to Content
Merck
All Photos(1)

Key Documents

C9867

Sigma-Aldrich

CGK733

≥98% (HPLC), solid

Synonym(s):

2,2-Diphenyl-n-(2,2,2-trichloro-1-[3-(4-fluoro-3-nitrophenyl)thioureido]ethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18Cl3FN4O3S
CAS Number:
Molecular Weight:
555.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cc(NC(=S)NC(NC(=O)C(c2ccccc2)c3ccccc3)C(Cl)(Cl)Cl)ccc1F

InChI

1S/C23H18Cl3FN4O3S/c24-23(25,26)21(30-22(35)28-16-11-12-17(27)18(13-16)31(33)34)29-20(32)19(14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-13,19,21H,(H,29,32)(H2,28,30,35)

InChI key

HLCDNLNLQNYZTK-UHFFFAOYSA-N

Biochem/physiol Actions

Originally published as a selective ataxia telangiectasia-mutated (ATM) and ATM- and Rad3-related (ATR) kinase inhibitor, which causes reversible reprogramming of cellular lifespan, conferred robust growth to senescent cells that had ceased proliferation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga V Leontieva et al.
Aging, 3(11), 1078-1091 (2011-12-14)
In yeast, chronological senescence (CS) is defined as loss of viability in stationary culture. Although its relevance to the organismal aging remained unclear, yeast CS was one of the most fruitful models in aging research. Here we described a mammalian
Ling Hui Xu et al.
The Journal of biological chemistry, 286(45), 39546-39559 (2011-09-16)
Perturbation of cell cycle regulation is a characteristic feature of infection by many DNA and RNA viruses, including Coronavirus infectious bronchitis virus (IBV). IBV infection was shown to induce cell cycle arrest at both S and G(2)/M phases for the
Yi Na Jiang et al.
Experimental cell research, 381(2), 165-171 (2019-05-18)
Matrix metalloproteinases (MMPs) and the epithelial-mesenchymal transition (EMT) contribute to metastasis. As shown in our previous studies, interleukin-6 (IL-6) induces ATM phosphorylation to increase MMP expression and metastasis in lung cancer. However, the exact roles of ATM activation in the
Huan Wang et al.
Biochemical and biophysical research communications, 422(1), 103-108 (2012-05-09)
Hepatocellular carcinoma (HCC) is one of the most deadly human cancers. Chronic hepatitis B virus (HBV) infection is one of the predominant risk factors associated with the development of HCC and complicates the treatment of HCC. In this study, we
John P Alao et al.
Radiation oncology (London, England), 4, 51-51 (2009-11-12)
The ataxia telangiectasia mutated (ATM) and the ATM- related (ATR) kinases play a central role in facilitating the resistance of cancer cells to genotoxic treatment regimens. The components of the ATM and ATR regulated signaling pathways thus provide attractive pharmacological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service